missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Progesterone R/NR3C3 Antibody (PGR/8099R), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-23941-100ug
This item is not returnable.
View return policy
Description
Progesterone R/NR3C3 Monoclonal antibody specifically detects Progesterone R/NR3C3 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Progesterone R/NR3C3 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| NR3C3PRNuclear receptor subfamily 3 group C member 3, progesterone receptor | |
| Recombinant fragment (around aa 1-200) of human Progesterone R/NR3C3 (exact sequence is proprietary) | |
| 100 μg | |
| Breast Cancer, Cancer, Immunology, Innate Immunity, Phospho Specific, Signal Transduction | |
| 5241 | |
| Store at 4°C. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| PGR/8099R | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction