missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell™ proFood™ 79-340 Series Green Nitrile Gloves
Reusable gloves suitable for contact with all fatty foodstuffs.
Brand: Ansell™ 79340080
Code : NEW
Additional Details : Weight : 0.53100kg
Description
- Good mechanical and chemical properties
- Contains no natural rubber
- Safe for both food and workers hands
- Flexibility, even in cold condition
Applications:
- Poultry processing
- Beef and pig processing
- Fish processing
- Janitorial use
Specifications
Nitrile | |
Gloves | |
79-340 | |
Green | |
Cotton Flocking | |
Mechanical and chemical |
Nitrile | |
8 | |
0.3 mm | |
Straight | |
12 Pairs | |
79-340 |