Learn More
Nabertherm™ High-Temperature Furnaces with Molybdenum Disilicide Heating Elements and Fiber Insulation
Due to their solid construction and compact stand-alone design, these high-temperature furnaces are perfect for processes in the laboratory where the highest precision is needed. Oustanding temperature uniformity and practical details set very high quality benchmarks.
Brand:  Nabertherm™ HT0357VNA
Additional Details : Weight : 306.00000kg
 View more versions of this product
Product Code. 17387839
  • £21942.43 / Each
Estimated Shipment: 25-07-2024
to see stock.
Add to basket
Description
Description
  • Dual shell housing with fan cooling for low shell temperatures
  • Heating from both sides via molybdenum disilicide heating elements
  • High-quality fiber insulation backed by special insulation
  • Side insulation constructed with tongue and groove blocks provides for low heat loss to the outside
  • Long-life roof insulation with special suspension
  • Chain-guided parallel swivel door for defined opening and closing of the door
  • Vapor vent in the furnace roof with motorized exhaust air flaps, controlled via the extra function of the controller
  • Heating elements switched via thyristors
Specifications
Specifications
P570
Up to 1750°C
300 x 200 x 260 mm
400 V
16 L
Molybdenum Disilicide Heating Elements and Fiber Insulation
690 x 820 x 1860 mm
12500 W