missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek Cytation 1 Cell Imaging Multi-Mode Reader

BioTek Cytation 1 cell imaging multi-mode reader combines fluorescence and high contrast brightfield imaging with conventional multi-mode detection. For Research Use Only. Not for use in diagnostic procedures.

Brand:  Agilent BioTek CYT1FAV-SN


Additional Details : Weight : 45.00000kg

 View more versions of this product

Product Code. 17247929

  • POA / Each
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.


Description

Description

BioTek Cytation 1 cell imaging multi-mode reader combines fluorescence and high contrast brightfield imaging with conventional multi-mode detection. This affordable imaging reader delivers performance and economy compared to other digital microscopy systems

The BioTek Cytation 1 design provides both quantitative phenotypic cellular data with well-based quantitative data. The multi-mode detection module has high sensitivity filter-based fluorescence and a monochromator for UV-Vis absorbance. Temperature control and shaking are standard. CO2/O2 control and reagent injectors are available. Gen5 software simplifies image capture and plate reading.

Specifications

Specifications

Cell Imaging Multi-Mode Microplate Reader
0 to 20%
Fluorescence and high contrast brightfield imaging; Monochromator absorbance; Filter/dichroic fluorescence; Luminescence/filtered luminescence
6-well to 384-well plates
Xenon Flash
Gen5 Microplate Reader and Imager Software
20 in.
17.5 in.
16.5 in.
CE and TUV marked. RoHS compliant.
Linear, Orbital, Double Orbital
0 to 19%
CYT1FAV
Endpoint, kinetic, area scanning, plate mode inject/read process and well mode inject/read process
0.3 μm/pixel at 20X
Incubation to 45°C
50.8 cm
44.5 cm
41.9 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers