missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ E-Gel™ Power Snap Electrophoresis Device Starter Kit, EX DC 1%

The Invitrogen™ E-Gel™ Power Snap Electrophoresis Device Starter Kit, EX DC 1%, provides everything you need to quickly start using precast E-Gel™ EX Double Comb 1% agarose gels on the device.

Brand:  Invitrogen™ G8131ST


1208.42 GBP valid until 2024-06-28
Use promo code "21649" to get your promotional price.



Additional Details : Weight : 2.30000kg

Product Code. 17165979

  • £1702.00 / Each
Estimated Shipment: 18-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The kit includes:

  • Ten E-Gel™ EX Double Comb Agarose Gels, 1% (Cat. No. A42345)
  • E-Gel™ Power Snap Electrophoresis Device (Cat. No. G8100)
  • E-Gel™ 1-Kb Plus Express DNA Ladder with E-Gel™ Sample Loading Buffer (1X) (Cat. No. 10488091)
Specifications

Specifications

• Ten E-Gel EX Double Comb Agarose Gels, 1% (Cat. No. A42345)
• E-Gel Power Snap Electrophoresis Device (Cat. No. G8100)
• E-Gel 1-Kb Plus Express DNA Ladder with E-Gel Sample Loading Buffer (1X) (Cat. No. 10488091)

Store at room temperature.
E-Base™, E-Gel™ Imager
Mini
Electrophoresis System Starter Kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.