missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ E-Gel™ Power Snap Electrophoresis System Starter Kit, EX DC 2%

The Invitrogen™ E-Gel™ Power Snap Electrophoresis System Starter Kit, EX DC 2%, provides everything you need to quickly start using precast E-Gel™ EX Double Comb 1% agarose gels on the system for a streamlined gel electrophoresis and gel imaging workflow.

Brand:  Invitrogen™ G8332ST


Additional Details : Weight : 3.80000kg

Product Code. 17115989

  • £2990.00 / Each
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The kit includes:

  • Ten E-Gel™ EX Double Comb Agarose Gels, 2% (Cat. No. A42346)
  • E-Gel™ Power Snap Electrophoresis System (Cat. No. G8300)
  • E-Gel™ 50-bp DNA Ladder with E-Gel™ Sample Loading Buffer (1X) (Cat. No. 10488099)
Specifications

Specifications

• Ten E-Gel EX Double Comb Agarose Gels, 2% (Cat. No. A42346)
• E-Gel Power Snap Electrophoresis System (Cat. No. G8300)
• E-Gel 50-bp DNA Ladder with E-Gel Sample Loading Buffer (1X) (Cat. No. 10488099)

Store at room temperature.
E-Base
Electrophoresis System Starter Kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.