Learn More
Applied Biosystems™ Automated Thermal Cycler (ATC) + Plastics Packages, 96-well
For customers seeking to automate their PCR workflow, the Automated Thermal Cycler (ATC) provides the reliability and performance you’ve come to expect from Applied Biosystems instruments.
Brand: Applied Biosystems™ A41198
Description
For customers seeking to automate their PCR workflow, the Automated Thermal Cycler (ATC) provides the reliability and performance you’ve come to expect from Applied Biosystems instruments. Now in a small, easy-to-integrate, and compatible format, our Automated Thermal Cycler enables hands-free PCR results.
This Automated Thermal Cycler (ATC) package includes a thermal cycler plus MicroAmp EnduraPlates (full-skirt or half-skirt) and MicroAmp Clear Adhesive Films.
Automated Thermal Cycler
• Space-saving design with our smallest thermal cycler footprint
• Flexible modular design helps save space on a robotic deck
• Separate control panel module for flexibility when integrating on a robotic deck
• Choice of 10-cm, 1-m, and 3-m cables for flexibility to locate control module under thermal cycler, under deck, or up to 3 meters away
• Automated lid compatible with programming for easy, hands-free operation with a liquid handler or plate stacker
• Heated cover slides forward to cover plate nest to help ensure thermal contact and eliminate evaporation
• Compatible with full-skirt or half-skirt plates
MicroAmp EnduraPlates
MicroAmp EnduraPlates offer excellent PCR or qPCR performance in formats developed to meet your experimental needs. They are a solution for work that requires special handling and an even greater degree of durability for use with multi-instrument experiments. They are validated for use with Applied Biosystems PCR thermal cyclers for optimal fit and performance.
• Optimized for performance with Applied Biosystems instruments
• Certified DNA-, RNAse-, and PCR inhibitor-free with in-process sampling tests
• Constructed to ANSI/SBS standard, well suited to high-throughput laboratories and robot handling
• Available in multiple colors for easy organization of laboratory assays
• Excellent mechanical stability and flatness for your complete workflow needs
MicroAmp Clear Adhesive Films
MicroAmp Clear Adhesive Films create a secure seal across an entire microplate to prevent evaporation and well-to-well contamination.
• Adhesive provides tight seal over wells to prevent leaks and evaporation.
• Easy to apply and remove
• Available as Clear (economical and sticky) or Optical (ready for real-time PCR and non-tacky)
Specifications
±0.25°C (35°C to 99.9°C) | |
High-throughput Compatible | |
Standard | |
96-well, 0.2 mL, half-skirted plate | |
No Display | |
Limited Warranty | |
USB, Wi-Fi, Ethernet | |
3.5°C/sec. (Block), 1.8°C/sec. (Sample) | |
13.2 lb. (6.0 kg) |
• Automated Thermal Cycler (ATC), 96-well, 1-meter cable • 5 x MicroAmp EnduraPlate Optical 96-well Clear Reaction Plates with Barcode (100 plates total) • 1 x MicroAmp Clear Adhesive Films (100 films) |
|
<0.5°C (20 sec. after reaching 95°C) | |
12.5 x 7.1 x 5.3 in. (31.7 x 17.9 x 13.3 cm) | |
100/240 V | |
Thermal Cycler | |
4°C to 105°C | |
600 W | |
Includes 5 x MicroAmp EnduraPlate Optical 96-well Clear Reaction Plates with Barcode (100 plates total) | |
96-well Isothermal Block |