Learn More
Sartorius arium™ comfort II Combined System (TYPE 1 - Ultrapure Water and TYPE 2 - EDI Water)
Use the Sartorius arium™ comfort I Combined System for producing ASTM Type 1 ultrapure water and Type 2 pure water.
Brand:  Sartorius H2O-II-1-TOC-T
Additional Details : Weight : 28.00000kg
Product Code. 16596374
  • £13590.00 / Each
Estimated Shipment: 26-06-2024
to see stock.
Add to basket
Description
Description
  • Flexible, space-saving twin technology
  • Efficient iJust technology
  • Uses disposable bagtank technology with dispense gun connections
  • Built-in TOC monitor analyzes and displays actual TOC values
  • Integrated UV lamp, 185/254nm, reduce the TOC less-than-or-equal-to 2ppb
  • Convenient SD card slot
  • User-friendly menu navigation
  • Meet customerapostrophes lab requirements and saves maintenance time
  • Safe operation
  • Reliable TOC reduction
  • Supports reliable recording of important data
To install arium comfort II as a built-in unit, please order the corresponding bench-top system and a conversion kit H2O-ACK-D
Specifications
Specifications
0.055 Product water, Less than1500 feed water
120 (ASTM type 1), 10 (Type 2)
Type 1 and 2
<2 product water, <1000 feed water
35.6°F to 86°F
HPLC, AAS, Ion chromatography, Photometry, Microbiological media and reagents, Histology, ELISA, RIA, Buffers and pH solutions Feed water for laboratory devices, such as autoclaves, glassware washers, humidifiers, water baths
UV lamp, TOC monitor
4 to 10
35.6°F to 95°F
<4
17.12 in.
19.7 in.
18.74 in.
79.36 lb.
100/240 V 50/60 Hz
<1 CFU/1000 mL
2 L/min. (ASTM type I), 0.3 L/min. (type II)
18.2 Mohm.cm product water
Water Purification System
2°C to 30°C
80%
<1/mL
5°C to 45°C
2°C to 35°C
<1
43.5 cm
50.1 cm
47.6 cm
36 kg