missing translation for 'onlineSavingsMsg'
Learn More

Asecos™ Q-PEGASUS-90 Two-Door Yellow Safety Storage Cabinets

For the safe and approved storage of flammable hazardous substances in working areas.

Brand:  Asecos™ 3004204033622


Code : NEW

Additional Details : Weight : 343.00000kg

 View more versions of this product

Product Code. 15871408

  • £3497.47 / Each
Estimated Shipment: 07-10-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Extremely robust design with scratch-proof surface
  • Triple-hinged doors
  • Safety elements assembled outside the storage compartment for increased protection against corrosion
  • One-hand operation for double-door cabinets
  • Container remains in one hand whilst opening
  • Doors remain open in the serial door open arrest system, convenient and safe removal or storage of containers
  • Doors automatically close after approx. 60 seconds
  • Acoustic and visual signal before door closing
  • Modern bicolor design - choice of seven different door colors
  • Language-free labelling for optimum international usability
  • Doors lockable with cylinder lock, cabinets can be integrated by the customer in an existing master key system
  • Locking status indicator (red/green) signals the locking status
  • Integrated air ducts ready for connection (DN 75) to a technical exhaust system
  • Visual inspection of the exhaust air function in the exhaust grille (rotating fan impeller)
  • Available with different combinations of drawers, height-adjustable shelves and bottom collecting sumps for leakages
  • In accordance with EN 14470-1 and TRGS 510
  • Fire resistance of 90 minutes
  • Integrated transport base with removable, optional base cover
  • Easy alignment – adjusting aids to compensate for uneven floors
Specifications

Specifications

615 x 893 x 1953 mm
Yellow
Safety storage cabinet
Shelves 75 kg; Insert 75 kg; Sump 22 L
2
394.5 kg
3 x 75 kg shelves; 1 x 75 kg perforated insert; 1 x 22 L bottom collecting sump
Storage of flammable hazardous substances
Auto-close
EN 14470-1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers