Learn More
SI Analytics TitroLine 7500 KF Trace Module 5, Coulometric KF-Titrator
These titrators from SI-Analytics are fast, easy, and precise and feature a robust, reliable, high visible full color display, that can be easily viewed from a distance and extreme angles.
Brand:  SI Analytics 285221000
Additional Details : Weight : 6.00000kg
Product Code. 15574923
  • £5195.00 / Each
Estimated Shipment: 22-07-2024
to see stock.
Add to basket
Description
Description
  • The TitroLine™ 7500 KF is the volumetric generalist for a wide range of use and the TitroLine™ 7500 KF trace is the specialist for low water contents
  • Fast, easy and precise
  • With standard methods for different applications (titer determination, blank value…)
  • High visible full color display, that can be easily viewed from a distance and extreme angles
  • Storage of results via USB and LAN port (PDF and CSV -format) with online curve
Specifications
Specifications
TitroLine 7500 KF Trace Module 5, Coulometric KF-Titrator
TitroLine 7500 KF Trace Module 5, Coulometric KF-Titrator