missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ 4 ft. Purifier Stainless Steel Horizontal Clean Bench

Purifier™ Horizontal Clean Benches direct HEPA-filtered air over the work area to provide ISO Class 5 conditions and protect from particulate contamination.

Brand:  Labconco™ 3888405


Additional Details : Weight : 206.00000kg

Product Code. 15311788

  • £11000.00 / Each
Estimated Shipment: 13-09-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Purifier™ Stainless Steel Horizontal Clean Benches direct HEPA-filtered air horizontally over the work area to provide ISO Class 5 (formerly Class 100) conditions and protect your work from particulate contamination.
  • Applications for clean benches include plant tissue culture, media plate preparation, electronics inspection, medical device assembly and pharmacy drug preparation.
  • Because they do not provide protection to the user, they should not be used in conjunction with biohazardous material, toxins or radionuclides.
Specifications

Specifications

Horizontal Clean Bench
Stainless Steel
CE
86.4 cm
122 cm
230 V
50 in.
127 cm
HEPA
4 A
34 in.
48.4 in.
50/60 Hz
4 ft.
122 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers