Description
- Offers Apo correction from UV through IR ranges
- Parfocal for both visible and IR imaging
- Coverslip-corrected objective that is optimized for imaging through #1.5 coverslips
- Offers optical performance from visible light to near infrared light
- Meets needs across spectrum of magnifications and optical specifications
Specifications
Objective Lens | |
EVOS™ XL Core Imaging System, EVOS™ FL Color Imaging System, EVOS™ XL Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System | |
S-Apochromat | |
1 Ea. |
EVOS | |
Vessels, Slides, Cell Culture Dishes, Flasks, Microtiter Well Plates | |
100X |