missing translation for 'onlineSavingsMsg'
Learn More

Sartorius arium™ Comfort I Combined System (TYPE 1 - Ultrapure Water and TYPE 3 - RO Water)

Use the Sartorius arium™ Comfort I Combined System for producing ASTM Type 1 ultrapure water and Type 3 pure water.

Brand:  Sartorius H2O-I-2-TOC-T


Code : NEW

Additional Details : Weight : 30.00000kg

 View more versions of this product

Product Code. 13497058

  • £10300.00 / Each
Estimated Shipment: 02-09-2024
to see stock.
Add to basket

Description

Description

arium comfort I systems produce ASTM Type 1 ultrapure water and Type 3 pure water. The systems features the latest reverse osmosis technology, and a unique filter cartridge for producing ultrapure water of the highest quality, delivering consistently high-grade water quality at rates of up to 120l/h. The quality of this water even exceeds the ASTM Type 1 specifications. The system's integrated UV lamp (185/254 nm) prevents microbiological growth and reduces the TOC content to a minimum. The current TOC value is continuously checked by an integrated TOC monitor specially developed for this ultrapure water system and is continuously displayed.

  • Reliable TOC content of <2 ppb for reproducible results
  • Automatically optimised water consumption with iJust
  • Easy-to-use - unique glass display with touch function

Benefits:

  • Produces Type 3 and Type ASTM 1 water from tap water all in one unit; adapted to the customer's needs; the display will conveniently remain at eye level
  • Optimized water consumption
  • Meet customer's lab requirements and saves maintenance time
  • Safe operation
  • Reliable TOC reduction
  • Supports reliable recording of important data
  • Easy operation

  • HPLC
  • GC-MS, AAS, ICP-MS
  • Ion chromatography
  • TOC-Analysis
  • Photometry
  • Buffer and pH-solutions
  • Feed water for ultrapure water systems, rinsing glassware and laboratory vessels, feed water for humidifiers, autoclaves and dish washer

Specifications

Specifications

0.055 Product water, Less than1500 feed water
Greater than99%
120 (ASTM type 1), 16 (Type 3)
Type 1 to 3
<2 product water, <2000 feed water
35.6°F to 86°F
Feed water for ultrapure water system, rinsing Glassware and laboratory vessels, feed water for humidifiers, autoclaves and dish washer
TOC monitor
<1/mL
Adsorption by spherical activated Carbon, catalyst, reverse osmosis, deionization, UV radiation, optional final particle and sterile filtration
35.6°F to 95°F
<4
17.12 in.
19.7 in.
18.74 in.
68.3 lb.
100/240 V 50/60 Hz
50/60 Hz
Greater than99%
<1 CFU/1000 mL
Up to 2 L/min. (ASTM type 1), Less than 3 L/min. (Type 3)
18.2 Mohm.cm product water
Arium Comfort I Combined System
2°C to 30°C
80%
Benchtop
4 to 10
5°C to 45°C
2°C to 35°C
<1
43.5 cm
50.1 cm
47.6 cm
31 kg
100/240 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers