missing translation for 'onlineSavingsMsg'
Learn More

Jenway™ Model 7300 Visible Single Beam Spectrophotometer

Ideal for routine testing in clinical, veterinary, pharmaceutical and QC laboratories. Jenway™ Model 7300 Visible Single Beam Spectrophotometer uses icon-driven software and has improved navigation system for easy and intuitive usability.

Brand:  Jenway™ 83058-15


2910.58 GBP valid until 2024-12-31
Use promo code "21966" to get your promotional price.



Code : 50

Additional Details : Weight : 10.00000kg

Product Code. 13104594

  • £3030.65 / Each
Estimated Shipment: 30-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Icon driven software
  • Small footprint
  • Autologging capabilities
  • Press to read xenon lamp
  • Extensive range of accessories available
  • 3 year warranty
Specifications

Specifications

–300 to 9999
Tungsten Halogen
Visible Spectrophotometer
For Use In Education And General Qc Laboratories
Single Beam
3 Years
6 kg
400 mm
5 nm
320 to 1000 nm
220 mm
1 nm
±2 nm
275 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers