missing translation for 'onlineSavingsMsg'
Learn More

LMS™ Series 2 Model 240A Cooled Incubator, 420 L, ABS Plastic

Temperature Range: -10°C to +50°C

Brand:  LMS™ 240A


Code : 04

Additional Details : Weight : 92.00000kg

Product Code. 12823476

  • £10409.20 / Each
Estimated Shipment: 05-07-2024
to see stock.
Add to basket

Description

Description

  • Range of freestanding cooled incubators with a white stove enamel mild steel exterior, foam insulation and a white high impact ABS plastic interior
  • Reliable and accurate temperature control
  • Standard and programmable, single and dual temperature models available
  • Temperature range -10.0°C to +50.0°C, temperature variation with average workload ±0.5°C
  • Self-evaporating condensate drip tray
  • White plastic coated wire shelves
  • Energy efficient control system and eco-friendly
  • P.I.D. heating and cooling to ensure stable internal temperature
  • Pre-set safety cutout
  • Hermetically sealed refrigeration system
  • Door lock
  • Digital controls are easy to read and simple to use
  • Full range of factory-fitted optional extras available, including LED lighting, access porthole, alarm system, internal socket and chart recorder
  • Ideal for applications such as growth studies, B.O.D.tests, seed germination and vernalisation, tissue culture, enzymology, serum studies and immunology, sample storage or product shelf life tests
Specifications

Specifications

ISO 9001:2008
Freestanding incubator
ABS Plastic
640 mm x 550 mm
Stove Enameled Mild Steel
-10°C to +50°C
Digital
240A
CFC-free
±0.5°C
29.5 in.
750 mm
69.7 in.
1770 mm
15.7 in.
92 kg
230 V
Standard digital with Automatic Dual Temperature Cycling
420 L
Fan assisted air circulation via premixing chamber
Optional LED
5
Refrigerated Incubator
Foamed polyurethane
1
±0.1°C
2 Years
25.8 in.
655 mm
57.1 in.
1450 mm
400 mm
240 V, 50 Hz
50 Hz
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers