missing translation for 'onlineSavingsMsg'
Learn More

Interscience Scan 1200 Automatic Colony Counter

Scan 1200 is an HD automatic colony counter. It adapts itself to all media and counts in 1 click all colonies even the smallest ones. It ensures accuracy and excellent reproducibility.

Brand:  Interscience 437000


Code : 50

Additional Details : Weight : 9.40000kg

Product Code. 12618695

  • POA / Each
Estimated Shipment: 04-07-2024
to see stock.

This item requires a price update prior to being ordered.

Request a Price for this item here.


Description

Description

HD automatic colony counter & inhibition zone reader, delivered with software.
  • Counting on Petri dishes (55 - 90 mm)
  • Spiral™ plating
  • Color detection: 7 colors on the same dish
  • Inhibition zone measurement
  • Counting on PetriFilmTM, RIDATM Count, Compact DryTM, filtration membrane
Specifications

Specifications

Automatic Colony Counter
Minimum size of detected colony: 0.05 mm
Counting: automatic with manual control (Counting on surface, Spiral, circle mode plated Petri dishes, Counting on chromogenic agar, PetriFilmTM, MC-media PadsTM, Compact DryTM, Easy PlateTM, filtration membranes, Inhibition zone reader)
Stainless Steel
Scan 1200
Colony counter
Number of antibiotic paper disks: up to 7 antibiotics on a dia. 90 mm Petri dish,Reading time: 7 inhibition zone reading between 1 to 3 seconds
14.69 in.
10.35 in.
9.4 kg
100/240 V
21 CFR Part 11, ISO 7218 and AOAC 977.27
Counting on round Petri Dishes of dia. 55 - 90 mm,PetriFilmTM, MC-Media PadsTM, Compact DryTM, easyPlateTM, filtration membranes, Compatible with dataLinkTM or dataLinkTMpro, traceability system
3-year warranty after warranty card recording
Scan 1200 is an HD automatic colony counter. It adapts itself to all media and counts in 1 click all colonies even the smallest ones. It ensures accuracy and excellent reproducibility. Images and results are saved automatically. Scan 1200 is also an inhibition zone reader. Diameter of round Petri Dishes : dia. 55, 65 and 90mm
Gray
HD CCD color camera,HD japanese lens, Zoom x 28, Resolution: 1.2 megapixels
Data export to PDF report, jpg, png and bmp images, ExcelTM recountable session
USB
1
28.8 x 26.3 x 37.3 cm
11.34 in.
20 lb
50-60 Hz
Max power: 20 W
The Scan 1200 colony counter is used in the food, environmental, pharmaceutical, cosmetics, veterinary and public institutes research
1 Scan™ software, 1 15 V power cord,1 USB cable, 3 validation plates, 1 user manual
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers