missing translation for 'onlineSavingsMsg'
Learn More

Carbolite™ Furnace with type 20/40 Temperature Sensor

Brand:  Carbolite™ HTF1804-230SN+TMP-3216P1BASE


Code : A0

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12089109

  • £19405.00 / Each
Estimated Shipment: 02-07-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Compliance to BSEN 61010
4 L
For use in quality control, heat treatment and research of ceramic materials.
830 mm
565 mm
650 mm
230 V 50/60 Hz 4650 w
4650 w
3216P1 Controller
Furnace
190 x 140 x 140 mm
140 mm
140 mm
190 mm
230 V
50/60 Hz
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers