Learn More
EVOS™ EVOS™ 60X Objective, fluorite, LWD
Components manufactured to ensure publication-quality images
Brand:  EVOS™ AMEP4626
Code : NEW
Additional Details : Weight : 0.01000kg
Product Code. 11957275
  • £3360.00 / Each
Estimated Shipment: 10-05-2024
to see stock.
Add to basket
Description
Description
RMS-threaded objectives with 45mm parfocal distance
Specifications
Specifications
Objective Lens
EVOS™ FL Color Imaging System, EVOS™ XL Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System
Plan Fluorite
0.75
EVOS
Vessels, Slides, Cell Culture Dishes, Flasks, Microtiter Well Plates
60X
1 Ea.