missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ KingFisher™ Flex Purification System, KingFisher with 96 PCR head

Prepare samples efficiently from a variety of materials with Thermo Scientific™ KingFisher™ Flex Magnetic Particle Processors.

Brand:  Thermo Scientific™ 5400610


Code : 50

Additional Details : Weight : 14.28810kg

Product Code. 11849520

  • £49760.00 / Each
Estimated Shipment: 02-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Prepare samples efficiently from a variety of materials with the Thermo Scientific™ KingFisher™ Flex Purification System, which offers highly versatile, automated magnetic-particle processing for DNA/RNA, protein or cell purification from virtually any source. Using revolutionary magnetic particle separation technology, this system provides excellent reproduciblity and quality.

Find a flexible solution for different types of sample processing needs. With 24 magnet heads, customers can raise the processing volume up to 5 mL, resulting in increased yield of the purified product. For higher throughput needs, 96 samples can be processed with 96 magnet head.
  • Fully automated system yields high-speed purification of nucleic acids, proteins and cells.
  • High throughput system processes up to 96 samples in less than 20 minutes.
  • Open and flexible system lets the customer use any magnetic particle-based kit to meet the application demands.
  • Easy-to-use Thermo Scientific™ BindIt™ Software provides instrument control, protocol creation and modification.
  • Ready-made protocols for different type of applications are available.

Recommended for:

  • Genomics and proteomics
  • Target identification
  • Veterinary assays
  • Biomarker discovery
  • Quality control
  • Forensics

Specifications

Specifications

20 to 100 μL
96-well Plate
KingFisher 96 plate, PCR plate
START / PAUSE / STOP / OK / TURNTABLE ROTATING CLOCKWISE / TURNTABLE ROTATING COUNTERCLOCKWISE / four cursor keys / LCD
From +5°C above ambient temperature to +115°C
ca. >1 μm
23.6 in.
15 in.
62 lb.
26.8 in.
24 or 96
96 Samples
> 95%
Genomics and proteomics, target identification, veterinary assays, biomarker discovery, quality control, forensics
96 in one frame; Interchangeable magnet heads
24 or 96-well plates: KingFisher 24 Deep Well plate (200 to 5000 μL), Microtiter 96 Deep Well plate (50 to 1000 μL), KingFisher 96 plate (20 to 200 μL), PCR plate (20 to 100 μL)
60 cm
38 cm
28 kg
68 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Laboratory Use