missing translation for 'onlineSavingsMsg'
Learn More

Techne™ Hybrigene HB-3D Hybridization Incubator with 4 Large Tubes

Temperature Range: Ambient +10°C to 80°C

Brand:  Techne™ 39455-06


Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 11583859

  • £4077.54 / Each
Estimated Shipment: 18-09-2024
to see stock.
Add to basket

Description

Description

HB-3D Hybrigene
  • Temperature range from 10°C above ambient to 80°C
  • Can hold up to 16 mini or 4 large glass tubes
  • Flexibility at an affordable price, offering excellent temperature accuracy and uniformity
  • The Hybrigene is a compact, stackable alternative to the Hybridizer HB-1D - for when space is important
  • Stack up to 3 ovens, saving on valuable laboratory space
HB-1D Hybridizer
  • Compact, easy to use and provides complete protection from hybridization hazards
  • A temperature range of 10°C above ambient to 100°C
  • High capacity which can hold up to 24 mini tubes or 6 unique large tubes
  • Hybridizations can be performed with a minimal volume of 5ml of probe
  • Adjustable feet to enable accurate leveling
  • Unique double-glazed glass door; quiet and safe, providing durable protection
Specifications

Specifications

Ambient +10° to 80°C
4 large tubes FHB16
0.5 to 20RPM
355 mm
21 kg
230 V
50/60 Hz
Hybridization Incubator
HB-3D
<±1.0°C
383 mm
230 V, 50/60 Hz, 750 w
750 w
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers