missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ accumet™ Solid-State Half-Cell ISEs, Mercury-Free: Chloride
Shop All Fisherbrand Products
Click to view available options
Connector Types:
BNC
Unit Size:
Each
Description
For chloride determinations.
- pH range: 0-14
- Material: Epoxy
- Connector type: BNC
Specifications
Specifications
| For Use With (Application) | Determination of Chloride concentration in water/wastewater, soil, dairy and tomato based food products, meats, vegetables and in research with biomedical fluids |
| Material | Epoxy |
| Connector Types | BNC |
| Type | Solid-State Half-Cell ISEs |
| pH Range | 0 to 14 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction