missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ Stainless Steel OIML Class E1 Reference Line Weight Sets With DAkkS Calibration Certificate

Brand:  Sartorius™ YCS31-611-02


Code : NEW

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 10285551

  • £5940.00 / Each
Estimated Shipment: 09-07-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Wood
1 g to 1 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers