missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Procollagen I N-Peptide Monoclonal Antibody (H20), Invitrogen™

Mouse Monoclonal Antibody
£178.00 - £408.00
Specifications
| Antigen | Procollagen I N-Peptide |
|---|---|
| Clone | H20 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|
17694336
|
Invitrogen™
MA543885 |
20 μL |
£178.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17684336
![]() |
Invitrogen™
MA543884 |
100 μL |
£408.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
Procollagen type III N-terminal peptide (P3NP) is released as part of collagen synthesis process in muscle. The increase of circulating P3NP is a marker not only of muscle growth, but also of muscle repair and fibrosis. P3NP may be a potential biomarker for sarcopenia.Spezifikation
| Procollagen I N-Peptide | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P1NP; Procollagen I N | |
| COL1A1 | |
| IgG1 κ | |
| Protein A/G | |
| Antibody |
| H20 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Rat | |
| P02452 | |
| 1277 | |
| Recombinant PINP (Gln23-Pro161) expressed in E.coli. | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| COL1A1 |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts