missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Procollagen I N-Peptide Monoclonal Antibody (H20), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA543885
This item is not returnable.
View return policy
Description
Procollagen type I N-terminal propeptide (P1NP) is a specific marker of bone formation, reflecting the synthesis of type I collagen, which constitutes the major part of the bone matrix. P1NP is derived during the conversion of procollagen to collagen, produced by osteoblasts during bone formation. Its primary clinical application lies in the diagnosis and monitoring of metabolic bone diseases, particularly osteoporosis. In osteoporotic patients, P1NP levels can predict bone mineral density (BMD) responses to treatments like teriparatide and romosozumab. Low serum levels of P1NP are indicative of a high risk of BMD loss, providing a valuable tool for premenopausal women with systemic lupus erythematosus (SLE) and other conditions affecting bone health.
Specifications
| Procollagen I N-Peptide | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P02452 | |
| COL1A1 | |
| Recombinant PINP (Gln23-Pro161) expressed in E.coli. | |
| 20 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG1 κ |
| Western Blot | |
| H20 | |
| Unconjugated | |
| COL1A1 | |
| P1NP; Procollagen I N | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 1277 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction