missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ProBond™ Nickel-Chelating Resin

Product Code. p-7182886
Click to view available options
Quantity:
50 mL
150 mL
Unit Size:
150mL
50mL
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11500656

Brand: Invitrogen™ R80115

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

A nickel-charged affinity resin used to purify recombinant proteins containing a polyhistidine (6xHis) sequence.

  • ProBond™ Nickel-Chelating Resin is a nickel-charged affinity resin used to purify recombinant proteins containing a polyhistidine (6xHis) sequence
  • Proteins bound to the resin may be eluted with either low pH buffer or by competition with imidazole or histidine
  • One-step purification can be performed under both native and denaturing conditions
  • ProBond™ Resin uses the chelating ligand iminodiacetic acid (IDA) coupled to a highly cross-linked 6% agarose resin
  • Suitable for use in FPLC, batch, and gravity flow applications
  • Guaranteed stable for 6 months when properly stored at 4°C
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage The ProBond™ resin is pre-charged and able to bind 1-5 mg of recombinant protein per 1 ml of resin. It is provided as a 50% slurry in 20% ethanol. The resin will appear blue in color when charged with Ni2+ . Store at +4°C. ProBond™ resin is guaranteed stable for 6 months when properly stored.
Stationary Phase Nickel-Chelating
Product Line ProBond
Type Resin
Quantity 150 mL
Form Suspension
Column Type Affinity Column

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.