missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRMT1 Antibody (PRMT1-171) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£175.00
Specifications
| Antigen | PRMT1 |
|---|---|
| Clone | PRMT1-171 |
| Concentration | 2 mg/mL |
| Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunoprecipitation 1:10-1:500 |
| Applications | Western Blot |
Description
PRMT1 Monoclonal specifically detects PRMT1 in Human, Mouse, Rat, Bovine, Canine, Chicken, Primate samples. It is validated for Western Blot, ELISA, Immunoprecipitation.Specifications
| PRMT1 | |
| 2 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Chromatin Research, Core ESC Like Genes, Signal Transduction, Stem Cell Markers | |
| ANM1, EC 2.1.1, EC 2.1.1.-, EC 2.1.1.125, EC 2.1.1.77, HCP1, Histone-arginine N-methyltransferase PRMT1, HMT1 (hnRNP methyltransferase, S. cerevisiae)-like 2, HMT1 hnRNP methyltransferase-like 2 (S. cerevisiae), HMT2, HRMT1L2HMT1 hnRNP methyltransferase-like 2, Interferon receptor 1-bound protein 4, IR1B4protein arginine N-methyltransferase 1, protein arginine methyltransferase 1 | |
| PRMT1 | |
| IgG1 | |
| Protein A or G purified | |
| This reacts with PRMT1 and does not cross-react with PRMT2, 3, 4, and 5. |
| PRMT1-171 | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunoprecipitation 1:10-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 3276 | |
| Monoclonal Anti-PRMT1 is derived from the PRMT1-171 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with full length PRMT1. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title