missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRMT1 Antibody (PRMT1-171) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB120-12189-0.025ml
This item is not returnable.
View return policy
Description
PRMT1 Monoclonal specifically detects PRMT1 in Human, Mouse, Rat, Bovine, Canine, Chicken, Primate samples. It is validated for Western Blot, ELISA, Immunoprecipitation.
Specifications
| PRMT1 | |
| Monoclonal | |
| 2 mg/mL | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunoprecipitation 1:10-1:500 | |
| PRMT1 | |
| Monoclonal Anti-PRMT1 is derived from the PRMT1-171 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with full length PRMT1. | |
| 0.025 mL | |
| Chromatin Research, Core ESC Like Genes, Signal Transduction, Stem Cell Markers | |
| 3276 | |
| Human | |
| Purified |
| Western Blot | |
| PRMT1-171 | |
| Unconjugated | |
| ANM1, EC 2.1.1, EC 2.1.1.-, EC 2.1.1.125, EC 2.1.1.77, HCP1, Histone-arginine N-methyltransferase PRMT1, HMT1 (hnRNP methyltransferase, S. cerevisiae)-like 2, HMT1 hnRNP methyltransferase-like 2 (S. cerevisiae), HMT2, HRMT1L2HMT1 hnRNP methyltransferase-like 2, Interferon receptor 1-bound protein 4, IR1B4protein arginine N-methyltransferase 1, protein arginine methyltransferase 1 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| This reacts with PRMT1 and does not cross-react with PRMT2, 3, 4, and 5. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction