missing translation for 'onlineSavingsMsg'
Learn More

Prilocaine hydrochloride, 98%, Thermo Scientific™

A local anesthetic

£41.65 - £181.00

Chemical Identifiers

CAS 1786-81-8
Molecular Formula C13H21ClN2O
Molecular Weight (g/mol) 256.774
MDL Number MFCD00079279
InChI Key BJPJNTKRKALCPP-UHFFFAOYSA-N
Synonym prilocaine hydrochloride, propitocaine hydrochloride, prilocaine hcl, xylonest, prilocaine chloride, citanest hydrochloride, citanest plain, n-2-methylphenyl-2-propylamino propanamide hydrochloride, 2-propylamino
PubChem CID 92163
ChEBI CHEBI:32053
IUPAC Name N-(2-methylphenyl)-2-(propylamino)propanamide;hydrochloride
SMILES CCCNC(C)C(=O)NC1=CC=CC=C1C.Cl
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15403389
SDS View Documents
Thermo Scientific Alfa Aesar
J60648.03
1g
ONLINE EXCLUSIVE PRICE
£45.60
£41.65 / 1g
Save £3.95
8% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15413389
SDS View Documents
Thermo Scientific Alfa Aesar
J60648.06
5g
£143.00
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15423389
SDS View Documents
Thermo Scientific Alfa Aesar
J60648.09
10g
£181.00
10g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1786-81-8
256.774
BJPJNTKRKALCPP-UHFFFAOYSA-N
92163
N-(2-methylphenyl)-2-(propylamino)propanamide;hydrochloride
C13H21ClN2O
MFCD00079279
prilocaine hydrochloride, propitocaine hydrochloride, prilocaine hcl, xylonest, prilocaine chloride, citanest hydrochloride, citanest plain, n-2-methylphenyl-2-propylamino propanamide hydrochloride, 2-propylamino
CHEBI:32053
CCCNC(C)C(=O)NC1=CC=CC=C1C.Cl

Specifications

1786-81-8
MFCD00079279
prilocaine hydrochloride, propitocaine hydrochloride, prilocaine hcl, xylonest, prilocaine chloride, citanest hydrochloride, citanest plain, n-2-methylphenyl-2-propylamino propanamide hydrochloride, 2-propylamino
CCCNC(C)C(=O)NC1=CC=CC=C1C.Cl
256.774
CHEBI:32053
98%
C13H21ClN2O
14,7743
BJPJNTKRKALCPP-UHFFFAOYSA-N
N-(2-methylphenyl)-2-(propylamino)propanamide;hydrochloride
92163
256.77
Prilocaine hydrochloride
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 217-244-0

missing translation for 'rtecsNumber' : UG5775000

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.