missing translation for 'onlineSavingsMsg'
Learn More

Prilocaine hydrochloride, 98%, Thermo Scientific™

A local anesthetic

Brand:  Thermo Scientific Alfa Aesar J60648.03

Quantity 1g

 View more versions of this product

Product Code. 15403389

  • ONLINE EXCLUSIVE PRICE
    £45.60
    £41.65 / 1g
    Save £3.95
    8% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

1786-81-8
256.774
BJPJNTKRKALCPP-UHFFFAOYSA-N
92163
N-(2-methylphenyl)-2-(propylamino)propanamide;hydrochloride
C13H21ClN2O
MFCD00079279
prilocaine hydrochloride, propitocaine hydrochloride, prilocaine hcl, xylonest, prilocaine chloride, citanest hydrochloride, citanest plain, n-2-methylphenyl-2-propylamino propanamide hydrochloride, 2-propylamino
CHEBI:32053
CCCNC(C)C(=O)NC1=CC=CC=C1C.Cl

Specifications

1786-81-8
MFCD00079279
14,7743
BJPJNTKRKALCPP-UHFFFAOYSA-N
N-(2-methylphenyl)-2-(propylamino)propanamide;hydrochloride
92163
256.77
Prilocaine hydrochloride
C13H21ClN2O
1g
prilocaine hydrochloride, propitocaine hydrochloride, prilocaine hcl, xylonest, prilocaine chloride, citanest hydrochloride, citanest plain, n-2-methylphenyl-2-propylamino propanamide hydrochloride, 2-propylamino
CCCNC(C)C(=O)NC1=CC=CC=C1C.Cl
256.774
CHEBI:32053
98%
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.