missing translation for 'onlineSavingsMsg'
Learn More

Kappes Systeme™ Premium Storage Bins

Product Code. p-8008285
Encompass_Preferred
Click to view available options
Color:
Blue
Gray
Green
Red
Yellow
Style:
Size 1
Size 2
Size 3
Size 4
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12652045

Brand: Kappes Systeme™ 6001.00.0150

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Type Storage Bin
Material Polyethylene
Color Blue
Shape Rectangular
For Use With Feed and Roller Conveyor
For Use With (Application) Food Storage
Style Size 1
Length (Metric) 500 mm
Width (Metric) 300 mm
Height (Metric) 250 mm
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.