missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Premium-Line Low Retention Pipette Tips
Shop All Fisherbrand Products
Click to view available options
Volume (Metric):
0.1 to 10 μL
0.5 to 20 μL
1 to 200 μL
100 to 1000 μL
Sterility:
Non-sterile
Sterile
Format:
Bulk
Rack
Unit Size:
Pack of 1000
Pack of 960
Description
- Optimized cone geometry provides a secure fit
- Graduation marks on tip at standard volumes
- Precise pipetting without loss of sample
- Conical tip opening prevents drop formation at the end of the tip
- Elimination of sample contamination
- Made of autoclavable, non-wettable, virgin polypropylene
- Tips are autoclavable fo sterile handling
Specifications
Specifications
| Volume (Metric) | 0.1 to 10 μL |
| Filtered | Non-filtered |
| Sterility | Non-sterile |
| Color | Clear |
| Autoclavable | Autoclavable |
| Length (Metric) | 34.1mm |
| Certifications/Compliance | CE, IVD |
| Sterilization Method | Autoclave |
| Format | Rack |
| Graduated | Yes |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction