missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRDM15 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00 - £470.00
Specifications
| Antigen | PRDM15 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18286361
|
Novus Biologicals
NBP2-58475 |
100 μL |
£470.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18648168
|
Novus Biologicals
NBP2-58475-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PRDM15 Polyclonal specifically detects PRDM15 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| PRDM15 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| 63977 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:DKPMLKQAGSGVHAAGTPENSAPVESEPSQWACKVCSATFLELQLLNEHLLGHLEQAKSLPPGSQSEAAAPEKEQD | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| C21orf83, chromosome 21 open reading frame 83, PFM15, PR domain containing 15, PR domain zinc finger protein 15, PR domain-containing protein 15, Zinc finger protein 298, ZNF298 | |
| PRDM15 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title