missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRCC Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | PRCC |
|---|---|
| Concentration | 0.24 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16819483
|
Proteintech
12019-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16809483
|
Proteintech
12019-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PRCC may regulate cell cycle progression through interaction with MAD2L2.Specifications
| PRCC | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q92733 | |
| 5546 | |
| PRCC Fusion Protein Ag2638 | |
| Primary | |
| -20°C | |
| PRCC |
| 0.24 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| PRCC, Proline rich protein PRCC, RCCP1, TPRC | |
| PRCC | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title