missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRCC Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12019-1-AP-150UL
This item is not returnable.
View return policy
Description
PRCC may regulate cell cycle progression through interaction with MAD2L2.Specifications
| PRCC | |
| Polyclonal | |
| Unconjugated | |
| PRCC | |
| PRCC, Proline rich protein PRCC, RCCP1, TPRC | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 5546 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.24 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q92733 | |
| PRCC | |
| PRCC Fusion Protein Ag2638 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction