missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRAF1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-56449
This item is not returnable.
View return policy
Description
PRAF1 Polyclonal specifically detects PRAF1 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| PRAF1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| DNA-directed RNA polymerase I subunit E, DNA-directed RNA polymerase I subunit RPA49, FLJ13390, FLJ13970, FLJ43482, PAF53RNA polymerase I-associated factor 1, polymerase (RNA) I associated factor 1, polymerase (RNA) I polypeptide E, 53kDa, PRAF1, RNA polymerase I associated factor 53, RNA polymerase I subunit A49, RNA polymerase I-associated factor 53, RP11-405L18.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 64425 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q9GZS1-2 | |
| POLR1E | |
| Synthetic peptides corresponding to POLR1E (polymerase (RNA) I polypeptide E, 53kDa) The peptide sequence was selected from the N terminal of POLR1E)(50ug). Peptide sequence SPGNMRFTLYENKDSTNPRKRNQRILAAETDRLSYVGNNFGTGALKCNTL. | |
| 100 μL | |
| Primary | |
| This product is specific to Subunit or Isoform: RPA49. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction