missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PR Mouse anti-Human, Clone: 3D1D12, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | PR |
|---|---|
| Clone | 3D1D12 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16898045
|
Proteintech
66300-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16888045
|
Proteintech
66300-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The progesterone receptor is a member of the Steroid Hormone Receptor Superfamily. It exists in two distinct isoforms in human cells. PR-B is the transcriptionally active form and is responsible for activating genes for the maintainence of the endometrium, maintece of pregcy, and inhibition of ovulation. PR-A is identical to PR-B except for a 165 amino acid deletion at the N-terminus. This deletion exposes a 140 amino acid inhibitory domain (ID) that acts as a repressor of steroid hormone transcriptional activity. Phospholabeling studies have shown that PR is phosphorylated in at least 9 different locations in response to progestins, which suggests that PR affects many different cellular pathways.Specifications
| PR | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| NR3C3, PGR, PR, progesterone receptor | |
| PGR | |
| IgG1 | |
| Protein G | |
| Antibody |
| 3D1D12 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human | |
| P06401 | |
| 5241 | |
| PR Fusion Protein Ag22992 | |
| Primary | |
| -20°C | |
| PGR |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title