missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PR Mouse anti-Human, Clone: 3D1D12, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66300-1-IG-150UL
This item is not returnable.
View return policy
Description
The progesterone receptor is a member of the Steroid Hormone Receptor Superfamily. It exists in two distinct isoforms in human cells. PR-B is the transcriptionally active form and is responsible for activating genes for the maintainence of the endometrium, maintece of pregcy, and inhibition of ovulation. PR-A is identical to PR-B except for a 165 amino acid deletion at the N-terminus. This deletion exposes a 140 amino acid inhibitory domain (ID) that acts as a repressor of steroid hormone transcriptional activity. Phospholabeling studies have shown that PR is phosphorylated in at least 9 different locations in response to progestins, which suggests that PR affects many different cellular pathways.Specifications
| PR | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P06401 | |
| PGR | |
| PR Fusion Protein Ag22992 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin) | |
| 3D1D12 | |
| Unconjugated | |
| PGR | |
| NR3C3, PGR, PR, progesterone receptor | |
| Mouse | |
| Protein G | |
| RUO | |
| 5241 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction