missing translation for 'onlineSavingsMsg'
Learn More

Cowie Technology™ Polypropylene Stir Bar Retriever

Product Code. 16225871 Shop All Fisherbrand Products
Click to view available options
Length (Metric):
350 mm
450 mm
Unit Size:
Pack of 5

Product Code. 16225871

Only null left
Add to basket
Add to basket

Use to retrieve stir bars from mixing containers. The polypropylene stir bar retriever is chemically resistant, inert, and leak-free. Uses Alnico V magnets at its core.

  • Inert and leak-free
  • Molded in pure, FDA-approved polypropylene
  • Alnico V magnets
  • Autoclavable

Specifications

Color White
Description Polypropylene encapsulated Alnico V magnet core
For Use With (Application) Removal of Stirrer Bars from vessels
For Use With (Equipment) PTFE Stirrer Bars
Diameter (Metric) 10 mm
Length (Metric) 450 mm
Quantity 1/Ea.
Type Polypropylene Stirrer Bar Retriever
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.