missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PPP6C Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£162.00 - £386.00
Specifications
| Antigen | PPP6C |
|---|---|
| Dilution | Western Blot : 1-10 μg/mL, Immunohistochemistry, Immunoprecipitation : 10-20 μg/mL |
| Applications | Immunohistochemistry, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
|
30067196
|
Novus Biologicals™
NBP3-30786-100ug |
100 μg |
£386.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30067040
|
Novus Biologicals™
NBP3-30786-25ug |
25 μg |
£162.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PPP6C Polyclonal antibody specifically detects PPP6C in Bovine,Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western BlotSpécification
| PPP6C | |
| Immunohistochemistry, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication, Signal Transduction | |
| Phosphate buffered saline | |
| 5537 | |
| IgG | |
| Ammonium sulfate precipitation |
| Western Blot : 1-10 μg/mL, Immunohistochemistry, Immunoprecipitation : 10-20 μg/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| Bovine, Human, Mouse, Rat | |
| EC 3.1.3.16, FLJ92648, MGC12249, PP6, PP6C, PPP6, protein phosphatase 6, catalytic subunit, serine/threonine protein phosphatase catalytic subunit, serine/threonine-protein phosphatase 6 catalytic subunit | |
| The immunogen used to generate the purified antibody was a peptide conjugated to KLH corresponding to the sequence NH2-Cys-Ala-Val-Pro-Asp-Ser-Glu-Arg-Val-IlePro-Pro-Arg-Thr-Thr-Thr-Pro-Tyr-COOH. This peptide antibody corresponds to C -terminal peptide of PPP6C catalytic subunit having a MW of 33kDa. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit