missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PPP6C Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30786-100ug
This item is not returnable.
View return policy
Description
PPP6C Polyclonal antibody specifically detects PPP6C in Bovine,Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot
Specifications
| PPP6C | |
| Polyclonal | |
| Western Blot : 1-10 μg/mL, Immunohistochemistry, Immunoprecipitation : 10-20 μg/mL | |
| EC 3.1.3.16, FLJ92648, MGC12249, PP6, PP6C, PPP6, protein phosphatase 6, catalytic subunit, serine/threonine protein phosphatase catalytic subunit, serine/threonine-protein phosphatase 6 catalytic subunit | |
| The immunogen used to generate the purified antibody was a peptide conjugated to KLH corresponding to the sequence NH2-Cys-Ala-Val-Pro-Asp-Ser-Glu-Arg-Val-IlePro-Pro-Arg-Thr-Thr-Thr-Pro-Tyr-COOH. This peptide antibody corresponds to C -terminal peptide of PPP6C catalytic subunit having a MW of 33kDa. | |
| 100 μg | |
| Cell Cycle and Replication, Signal Transduction | |
| 5537 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Phosphate buffered saline | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Bovine, Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction