missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP2R2B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£222.00
Specifications
| Antigen | PPP2R2B |
|---|---|
| Dilution | Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
PPP2R2B Polyclonal specifically detects PPP2R2B in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| PPP2R2B | |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| B55BETA, FLJ95686, MGC24888, PP2A subunit B isoform B55-beta, PP2A subunit B isoform beta, PP2A subunit B isoform PR55-beta, PP2A subunit B isoform R2-beta, PP2A, subunit B, B-beta isoform, PP2AB55BETA, PP2ABBETA, PP2APR55B, PP2APR55BETA, PR2AB55BETA, PR2ABBETA, PR2APR55BETA, PR52B, PR55BETA, PR55-BETA, protein phosphatase 2 (formerly 2A), regulatory subunit B (PR 52), beta isoform, protein phosphatase 2 (formerly 2A), regulatory subunit B, beta isoform, protein phosphatase 2, regulatory subunit B, beta, SCA12, serine/threonine protein phosphatase 2A, 55 kDa regulatory subunit B, betaisoform, serine/threonine protein phosphatase 2A, neuronal isoform, serine/threonine-protein phosphatase 2A 55 kDa regulatory subunit B betaisoform, spinocerebellar ataxia 12 | |
| PPP2R2B | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Polyclonal | |
| Rabbit | |
| Stem Cell Markers | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 5521 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:LPALHLQTSEHHPFFQLPHRRLGPWCSPTGSPAPLSCETGCGEGSWILVCRLLVPTQVSLLSMEEDIDTRKINN | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title