missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PPP1R14D Monoclonal Antibody (K49008_16C7), Invitrogen™
Mouse Monoclonal Antibody
£416.00 - £734.00
Specifications
| Antigen | PPP1R14D |
|---|---|
| Clone | K49008_16C7 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30196438
|
Invitrogen™
MA555913 |
50 μL |
£416.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30195008
|
Invitrogen™
MA556293 |
200 μL |
£734.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: MLSSSPASCT SPSPDGENPC KKVHWASGRR RTSSTDSESK SHPDSSKIPR SRRPSRLTVK YDRGQLQRWL EMEQWVDAQV QELFQDQATP SEPEIDLEAL MDLSTEEQKT QLEAILGNCP RPTEAFISEL LSQLKKLRRL SRPQK
PPP1R14D Monoclonal antibody specifically detects PPP1R14D in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| PPP1R14D | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| 2010107K19Rik; CPI17-like; gastrointestinal and brain-specific PP1-inhibitory protein 1; GBPI; GBPI1; GBPI-1; gut and brain phosphatase inhibitor 1; PKC-dependent PP1 inhibitory protein; PKC-dependent PP1 inhibitory protein subunit 14; Ppp1r14; PPP1R14D; protein phosphatase 1 regulatory inhibitor subunit 14D; protein phosphatase 1 regulatory subunit 14D; protein phosphatase 1, regulatory (inhibitor) subunit 14D | |
| Recombinant human PPP1R14D (immunogen range is 1-145). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| PPP1R14D |
| K49008_16C7 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9NXH3 | |
| 54866 | |
| IgG1 | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title