missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PPP1R14D Monoclonal Antibody (K49008_16C7), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA555913
This item is not returnable.
View return policy
Description
Sequence of this protein is as follows: MLSSSPASCT SPSPDGENPC KKVHWASGRR RTSSTDSESK SHPDSSKIPR SRRPSRLTVK YDRGQLQRWL EMEQWVDAQV QELFQDQATP SEPEIDLEAL MDLSTEEQKT QLEAILGNCP RPTEAFISEL LSQLKKLRRL SRPQK
Protein phosphatase-1 (PP1; see MIM 176875) is a major cellular phosphatase that reverses serine/threonine protein phosphorylation. PPP1R14D is a PP1 inhibitor that itself is regulated by phosphorylation (Liu et al., 2004 ).
Specifications
| PPP1R14D | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Q9NXH3 | |
| PPP1R14D | |
| Recombinant human PPP1R14D (immunogen range is 1-145). | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| K49008_16C7 | |
| Unconjugated | |
| PPP1R14D | |
| 2010107K19Rik; CPI17-like; gastrointestinal and brain-specific PP1-inhibitory protein 1; GBPI; GBPI1; GBPI-1; gut and brain phosphatase inhibitor 1; PKC-dependent PP1 inhibitory protein; PKC-dependent PP1 inhibitory protein subunit 14; Ppp1r14; PPP1R14D; protein phosphatase 1 regulatory inhibitor subunit 14D; protein phosphatase 1 regulatory subunit 14D; protein phosphatase 1, regulatory (inhibitor) subunit 14D | |
| Mouse | |
| Protein A | |
| RUO | |
| 54866 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction