missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PPP1R14D Monoclonal Antibody (K49008_16C7), Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA556293

 View more versions of this product

Product Code. 30195008

  • £734.00 / 200µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Sequence of this protein is as follows: MLSSSPASCT SPSPDGENPC KKVHWASGRR RTSSTDSESK SHPDSSKIPR SRRPSRLTVK YDRGQLQRWL EMEQWVDAQV QELFQDQATP SEPEIDLEAL MDLSTEEQKT QLEAILGNCP RPTEAFISEL LSQLKKLRRL SRPQK

Protein phosphatase-1 (PP1; see MIM 176875) is a major cellular phosphatase that reverses serine/threonine protein phosphorylation. PPP1R14D is a PP1 inhibitor that itself is regulated by phosphorylation (Liu et al., 2004 ).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PPP1R14D
Monoclonal
1 mg/mL
PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Q9NXH3
PPP1R14D
Recombinant human PPP1R14D (immunogen range is 1-145).
200 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
K49008_16C7
Unconjugated
PPP1R14D
2010107K19Rik; CPI17-like; gastrointestinal and brain-specific PP1-inhibitory protein 1; GBPI; GBPI1; GBPI-1; gut and brain phosphatase inhibitor 1; PKC-dependent PP1 inhibitory protein; PKC-dependent PP1 inhibitory protein subunit 14; Ppp1r14; PPP1R14D; protein phosphatase 1 regulatory inhibitor subunit 14D; protein phosphatase 1 regulatory subunit 14D; protein phosphatase 1, regulatory (inhibitor) subunit 14D
Mouse
Protein A
RUO
54866
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.