missing translation for 'onlineSavingsMsg'
Learn More

Lonza™ PPiLight™ Inorganic Pyrophosphate Assay, 500 Test

Product Code. 11600281
Change view
Click to view available options
Quantity:
500 Tests
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11600281 500 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11600281 Supplier Lonza™ Supplier No. LT07610
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rapid, efficient, luminescence-based pyrophosphate detection assay.

  • Fast- Measure enzyme activity via pyrophosphate consumption in 1 hour.
  • Simple - The 2-step, luminescent assay doesn't require radioactive substrates, beads, detection antibodies or modified substrates.
  • Wide Detection Range- Sensitive to 0.02 μM with a linear range to 10 μM.
  • Versatile - Scalable to 96-, 384-, 1586-well formats.
  • Applications:
    • Phosphotransferases
    • Pyrophosphotases
    • Cyclases
    • Ligases
    • Hydrolases
    • DNA Polymerases
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description 500 tests
For Use With (Application) Inorganic pyrophosphate assay
Includes 1 x LT27-263 PPiLight™ converting reagent, 10 mL, 1 x LT27-260 PPiLight™ detection reagent, 10 mL, 1 x LT27-266 PPiLight™ reconstitution buffer, 20 mL
Quantity 500 Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.