missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PPCS Recombinant Rabbit Monoclonal Antibody (24GB1350)
Rabbit Recombinant Monoclonal Antibody
£126.00 - £434.00
Specifications
| Antigen | PPCS |
|---|---|
| Clone | 24GB1350 |
| Concentration | 0.4 mg/mL |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30274995
|
Invitrogen™
MA557179 |
100 μL |
£434.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30276162
|
Invitrogen™
MA557180 |
20 μL |
£126.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PPCS Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Biosynthesis of coenzyme A (CoA) from pantothenic acid (vitamin B5) is an essential universal pathway in prokaryotes and eukaryotes. PPCS (EC 6.3.2.5), one of the last enzymes in this pathway, converts phosphopantothenate to phosphopantothenoylcysteine.Specifications
| PPCS | |
| 0.4 mg/mL | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9HAB8 | |
| 79717 | |
| A synthesized peptide derived from human PPCS (aa 290-311). | |
| Antibody |
| 24GB1350 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| PPCS | |
| 6330579B17Rik; AI413471; AI790434; Coab; Phosphopantothenate--cysteine ligase; phosphopantothenoylcysteine synthetase; PPC synthetase; Ppcs; RGD1311399 | |
| PPCS | |
| Primary | |
| Affinity chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title