missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PPCS Recombinant Rabbit Monoclonal Antibody (24GB1350)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557180
This item is not returnable.
View return policy
Description
PPCS Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Biosynthesis of coenzyme A (CoA) from pantothenic acid (vitamin B5) is an essential universal pathway in prokaryotes and eukaryotes. PPCS (EC 6.3.2.5), one of the last enzymes in this pathway, converts phosphopantothenate to phosphopantothenoylcysteine.
Specifications
| PPCS | |
| Recombinant Monoclonal | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9HAB8 | |
| PPCS | |
| A synthesized peptide derived from human PPCS (aa 290-311). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB1350 | |
| Unconjugated | |
| PPCS | |
| 6330579B17Rik; AI413471; AI790434; Coab; Phosphopantothenate--cysteine ligase; phosphopantothenoylcysteine synthetase; PPC synthetase; Ppcs; RGD1311399 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 79717 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction