missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PP4/PPP4C Antibody, Novus Biologicals™
Sheep Polyclonal Antibody
£162.00 - £386.00
Specifications
| Antigen | PP4/PPP4C |
|---|---|
| Dilution | Western Blot : 1:500-1:1000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence : 1:500-1:1000 |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30066823
|
Novus Biologicals™
NBP3-30787-100ug |
100 μg |
£386.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30067075
|
Novus Biologicals™
NBP3-30787-25ug |
25 μg |
£162.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PP4/PPP4C Polyclonal antibody specifically detects PP4/PPP4C in Bovine,Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunocytochemistry/ ImmunofluorescenceSpecifications
| PP4/PPP4C | |
| Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Sheep | |
| Core ESC Like Genes, Protein Phosphatase, Signal Transduction, Stem Cell Markers | |
| Phosphate buffered saline | |
| 5531 | |
| IgG | |
| Ammonium sulfate precipitation |
| Western Blot : 1:500-1:1000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence : 1:500-1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Bovine, Human, Mouse, Rat | |
| EC 3.1.3.16, PP4C, PP4protein phosphatase X, catalytic subunit, PP-X, PPXPPH3, protein phosphatase 4 (formerly X), catalytic subunit, protein phosphatase 4, catalytic subunit, Protein phosphatase X | |
| A PP X/C peptide conjugated to KLH | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title