missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PP4/PPP4C Antibody, Novus Biologicals™
Sheep Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30787-25ug
This item is not returnable.
View return policy
Description
PP4/PPP4C Polyclonal antibody specifically detects PP4/PPP4C in Bovine,Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| PP4/PPP4C | |
| Polyclonal | |
| Western Blot : 1:500-1:1000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence : 1:500-1:1000 | |
| EC 3.1.3.16, PP4C, PP4protein phosphatase X, catalytic subunit, PP-X, PPXPPH3, protein phosphatase 4 (formerly X), catalytic subunit, protein phosphatase 4, catalytic subunit, Protein phosphatase X | |
| A PP X/C peptide conjugated to KLH | |
| 25 μg | |
| Core ESC Like Genes, Protein Phosphatase, Signal Transduction, Stem Cell Markers | |
| 5531 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
| Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Phosphate buffered saline | |
| Sheep | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Bovine, Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction