missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PP2C beta/PPM1B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£162.00 - £386.00
Specifications
| Antigen | PP2C beta/PPM1B |
|---|---|
| Dilution | Western Blot : 5-10 μg/mL, Immunoprecipitation : 5-10 μg/mL |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30066789
|
Novus Biologicals™
NBP3-30782-25ug |
25 μg |
£162.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30067219
|
Novus Biologicals™
NBP3-30782-100ug |
100 μg |
£386.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PP2C beta/PPM1B Polyclonal antibody specifically detects PP2C beta/PPM1B in Bovine,Human,Mouse,Rat samples. It is validated for Immunoprecipitation,Western BlotSpecifications
| PP2C beta/PPM1B | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Stem Cell Markers | |
| Phosphate buffered saline | |
| 5495 | |
| IgG | |
| Ammonium sulfate precipitation |
| Western Blot : 5-10 μg/mL, Immunoprecipitation : 5-10 μg/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| Bovine, Human, Mouse, Rat | |
| EC 3.1.3.16, MGC21657, PP2C-beta, PP2C-beta-X, PP2CBmagnesium-dependent, beta isoform, protein phosphatase 1B, Protein phosphatase 2C isoform beta, protein phosphatase 2C-like protein, protein phosphatase, Mg2+/Mn2+ dependent, 1B | |
| The purified peptide conjugated to KLH. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title